Drukuj
Termostaty grzewcze, Alpha
Dostawca: LAUDA
L000619 L000620 L000621 L000623 L000624 L000625 L000627 L000628 L000629
461-1204EA 7640 PLN
461-1204 461-1205 461-1206 LAUDL000623 LAUDL000624 LAUDL000625 LAUDL000627 LAUDL000628 LAUDL000629
Termostaty grzewcze, Alpha
Termostaty cyrkulacyjne Systemy cyrkulacyjne, otwarte Łaźnie ze zdejmowanym cyrkulatorem
The Alpha heating thermostats have an easy to use control console featuring three button operation and large, clear LED display, ensuring easy navigation through the menu. The thermostats are suitable for use with non flammable liquids (water, water/glycol mixtures). Units can provide both internal and external thermostating. Temperature stability of ±0,05 K gives precise heating and cooling suitable for a wide range of applications. Using an optional pump circulation set and a cooling coil, the heating thermostat can be expanded to create a fully fledged thermostating system.
  • Temperature range 25 to 100 °C, resolution 0,1 °C, temperature stability of ±0,05 K
  • User-calibration at a single point for re-calibration of working temperatures
  • Pump flow rate can be reduced from 15 to 5 L/min by means of a reducer
  • Safety functions such as alarms, warnings and errors are shown on the LED display
Maximum pump capacities:
15 L/min, 200 mbar.
15 l/min, 200 mbar
Dodatkowe informacje: Supplied with stainless steel bath, flow reducer and screw clamp, which allows the immersion thermostat to be attached to any tank with a maximum wall thickness of 25 mm, and capacity of up to 50 litres; the maximum temperature achievable is dependent on the tank insulation.