Drukuj
384-dołkowe mikropłytki HTS do hodowli komórkowych, BRANDplates®
Dostawca: Brand
781980 781981 781982 781986 781987 781988 781989
735-2105EA 2340 PLN
735-2105 735-2106 735-2107 735-2111 735-2112 735-0372 735-0373
384-dołkowe mikropłytki HTS do hodowli komórkowych, BRANDplates®
Mikropłytki Płytki do hodowli komórkowych
PS, solid or with transparent base. These flat bottom plates are available in different colours, and with standard or transparent base formats.
cellGrade™: The standard surface for cultivation of adherent cell lines. Chemical groups on the surface of the plate bind to serum compounds, stimulating the growth of immobilised cells.
cellGrade™ plus: This surface has a protein like structure. Cultivation of cells in serum reduced media is possible. cellGrade™ plus surface is suitable for the cultivation of more fastidious cell lines.
cellGrade™ premium: This surface is a poly-D-lysineWięcej informacji o produkcie