Drukuj
Kulki szklane
Dostawca: Merck
18406-500G 18406-6X500G
SIAL18406-500GEA 304 PLN
SIAL18406-500G SIAL18406-6X500G
Kulki szklane
Kulki Kulki szklane
Made of PYREX® glass, solid.
For boiling and distillation operations. Use as distillation packing, mixing beads or boiling stones.