Drukuj
Biotyna-NHS
Dostawca: Merck
203118-50EA 1290 PLN
203118-50
Biotyna-NHS
Systemy do etykietowania i łączenia białek
Biotin-NHS reacts with primary amines to biotinylate proteins under mild conditions. Since it is water-soluble, this compound can be used to biotinylate cell-surface proteins and proteins that are sensitive to organic solvents.