Drukuj
Anti-NAPSA Mouse Monoclonal Antibody (CF405S) [clone: NAPSA/1239]
Dostawca: Biotium
BNC041239-500 BNC041239-100
BNC041239-500EA 2860 PLN
BNC041239-500 BNC041239-100
Anti-NAPSA Mouse Monoclonal Antibody (CF405S) [clone: NAPSA/1239]
Przeciwciała
Napsin is a pepsin-like aspartic proteinase connected with maturation of surfactant protein B.There are two closely related napsins, napsin A and napsin B. Napsin A is expressed as a single chain protein. Immunohistochemical studies revealed high expression levels of napsin A in human lung and kidney but low expression in spleen. Napsin A is expressed in type II pneumocytes and in adenocarcinomas of lung. The high specificity expression of napsin A in adenocarcinomas of lung is useful to distinguish primary lung adenocarcinomas from adenocarcinomas of other organs.
Type: Primary
Antigen: NAPSA
Clonality: Monoclonal
Clone: NAPSA/1239
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG kappa
Reactivity: Human