Drukuj

Anti-NAPSA Mouse Monoclonal Antibody (CF488A) [clone: NAPSA/1238]

Dostawca: Biotium
BNC881238-500 BNC881238-100
BNC881238-500EA 2860 PLN
BNC881238-500 BNC881238-100
Anti-NAPSA Mouse Monoclonal Antibody (CF488A) [clone: NAPSA/1238]
Przeciwciała
Napsin is a pepsin-like aspartic proteinase connected with maturation of surfactant protein B.There are two closely related napsins, napsin A and napsin B. Napsin A is expressed as a single chain protein. Immunohistochemical studies revealed high expression levels of napsin A in human lung and kidney but low expression in spleen. Napsin A is expressed in type II pneumocytes and in adenocarcinomas of lung. The high specificity expression of napsin A in adenocarcinomas of lung is useful to distinguish primary lung adenocarcinomas from adenocarcinomas of other organs.

Type: Primary
Antigen: NAPSA
Clonality: Monoclonal
Clone: NAPSA/1238
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR