Drukuj
Anti-CD2 Mouse Monoclonal Antibody (CF405S) [clone: UMCD2]
Dostawca: Biotium
BNC041235-500 BNC041235-100
BNC041235-500EA 2860 PLN
BNC041235-500 BNC041235-100
Anti-CD2 Mouse Monoclonal Antibody (CF405S) [clone: UMCD2]
Przeciwciała
CD2 interacts through its amino-terminal domain with the extracellular domain of CD58 (also designated CD2 ligand) to mediate cell adhesion. CD2/CD58 binding can enhance antigen-specific T cell activation. CD2 is a transmembrane glycoprotein that is expressed on peripheral blood T lymphocytes, NK cells and thymocytes. CD58 is a heavily glycosylated protein with a broad tissue distribution in hematopoietic and other cells, including endothelium. Interaction between CD2 and its counter receptor LFA3 (CD58) on opposing cells optimizes immune system recognition, thereby facilitating communication between helper T lymphocytes and antigen-presenting cells, as well as between cytolytic effectors and target cells.
Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: UMCD2
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human