Drukuj
Anti-TG Mouse Monoclonal Antibody (CF594) [clone: TGB04 TGB05]
Dostawca: Biotium
BNC941227-500 BNC941227-100
BNC941227-500EA 2860 PLN
BNC941227-500 BNC941227-100
Anti-TG Mouse Monoclonal Antibody (CF594) [clone: TGB04 TGB05]
Przeciwciała
Thyroglobulin is a 660 kDa dimeric pre-protein with mutiple glycosylation sites. It is produced by and processed within the thyroid gland to produce the hormone thyroxine and triiodothyronine. Prior to forming dimers, thyroglobulin monomers undergo conformational maturation in the endoplasmic reticulation. The vast majority of follicular carcinomas of the thyroid will give positive immunoreactivity for anti-thyroglobulin even though sometimes only focally. Poorly differentiated carcinomas of the thyroid are frequently anti-thyroglobulin negative. Adenocarcinomas of other-than-thyroid origin do not react with this antibody. This antibody is useful in identification of thyroid carcinoma of the papillary and follicular types. Presence of thyroglobulin in metastatic lesions establishes the thyroid origin of tumor. Anti-thyroglobulin, combined with anti-calcitonin, can identify medullary carcinomas of the thyroid. Furthermore, anti-thyroglobulin, combined with anti-TTF1, can be a reliable marker to differentiate between primary thyroid and lung neoplasms.
Type: Primary
Antigen: TG
Clonality: Monoclonal
Clone: TGB04 TGB05
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat