Drukuj

Anti-SUMO2 Mouse Monoclonal Antibody (CF640R) [clone: SUMO2/1199]

Dostawca: Biotium
BNC401199-500 BNC401199-100
BNC401199-500EA 2860 PLN
BNC401199-500 BNC401199-100
Anti-SUMO2 Mouse Monoclonal Antibody (CF640R) [clone: SUMO2/1199]
Przeciwciała
The small ubiquitin-related modifier (SUMO) proteins, which include SUMO-1, 2 and 3, belong to the ubiquitin-like protein family. Like ubiquitin, the SUMO proteins are synthesized as precursor proteins that undergo processing before conjugation to target proteins. Also, both utilize the E1, E2 and E3 cascade enzymes for conjugation. However, SUMO and ubiquitin differ with respect to targeting. Ubiquitination predominantly targets proteins for degradation, whereas sumoylation targets proteins to a variety of cellular processing, including nuclear transport, transcriptional regulation, apoptosis and protein stability. The unconjugated SUMO-1, 2 and 3 proteins localize to the nuclear membrane, nuclear bodies and cytoplasm, respectively. SUMO-1 utilizes Ubc9 for conjugation to several target proteins, which include MDM2, p53, PML and RanGap1. SUMO-2 and 3 contribute to a greater percentage of protein modification than does SUMO-1 and unlike SUMO-1, they can form polymeric chains. In addition, SUMO-3 regulates beta-Amyloid generation and may be critical in the onset or progression of Alzheimer s disease.

Type: Primary
Antigen: SUMO2
Clonality: Monoclonal
Clone: SUMO2/1199
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR