Drukuj
Anti-HSP90B1 Mouse Monoclonal Antibody (CF594) [clone: HSP90B1/1192]
Dostawca: Biotium
BNC941192-500 BNC941192-100
BTIUBNC941192-500EA 2860 PLN
BTIUBNC941192-500 BTIUBNC941192-100
Anti-HSP90B1 Mouse Monoclonal Antibody (CF594) [clone: HSP90B1/1192]
Przeciwciała
This antibody recognizes a protein of 94 kDa, which is identified as the glucose-regulated protein 94 (grp94) and also tumor rejection antigen (gp96). Grp94 shows a high degree of sequence homology with the heat shock protein 90 (hsp90). This MAb is highly specific to grp94 and shows minimal cross-reaction with other members of the HSP90 family. Grp s are a class of proteins unresponsive to heat shock and are induced by glucose deprivation. Grp94 has been briefly studied as a prognostic factor in breast cancer.
Type: Primary
Antigen: HSP90B1
Clonality: Monoclonal
Clone: HSP90B1/1192
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human