Drukuj

Anti-HSP90B1 Mouse Monoclonal Antibody (CF568) [clone: HSP90B1/1192]

Dostawca: Biotium
BNC681192-500 BNC681192-100
BTIUBNC681192-500EA 2860 PLN
BTIUBNC681192-500 BTIUBNC681192-100
Anti-HSP90B1 Mouse Monoclonal Antibody (CF568) [clone: HSP90B1/1192]
Przeciwciała
This antibody recognizes a protein of 94 kDa, which is identified as the glucose-regulated protein 94 (grp94) and also tumor rejection antigen (gp96). Grp94 shows a high degree of sequence homology with the heat shock protein 90 (hsp90). This MAb is highly specific to grp94 and shows minimal cross-reaction with other members of the HSP90 family. Grp s are a class of proteins unresponsive to heat shock and are induced by glucose deprivation. Grp94 has been briefly studied as a prognostic factor in breast cancer.

Type: Primary
Antigen: HSP90B1
Clonality: Monoclonal
Clone: HSP90B1/1192
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR