Drukuj

Anti-FBXO5 Mouse Monoclonal Antibody (CF405S) [clone: EMI1/1176]

Dostawca: Biotium
BNC041176-500 BNC041176-100
BNC041176-500EA 2860 PLN
BNC041176-500 BNC041176-100
Anti-FBXO5 Mouse Monoclonal Antibody (CF405S) [clone: EMI1/1176]
Przeciwciała
This antibody recognizes a 56 kDa protein, which is identified as Early Mitotic Inhibitor-1 (EMI1). It regulates mitosis by inhibiting the anaphase promoting complex/cyclosome (APC). Emi1 is a conserved F box protein containing a zinc-binding region essential for APC inhibition. The Emi1 protein functions to promote cyclin A accumulation and S phase entry in somatic cells by inhibiting the APC complex. At the G1-S transition, Emi1 is transcriptionally induced by the E2F transcription factor. Emi1 overexpression accelerates S-phase entry and can override a G1 block caused by overexpression of Cdh1 or the E2F-inhibitor p105 retinoblastoma protein (pRb). Depleting cells of Emi1 through RNA interference prevents accumulation of cyclin A and inhibits S phase entry.

Type: Primary
Antigen: FBXO5
Clonality: Monoclonal
Clone: EMI1/1176
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR