Drukuj
Anti-KRT19 Mouse Monoclonal Antibody (Biotin) [clone: KRT19/799 KRT19/800]
Dostawca: Biotium
BNCB1150-500 BNCB1150-100
BNCB1150-500EA 2860 PLN
BNCB1150-500 BNCB1150-100
Anti-KRT19 Mouse Monoclonal Antibody (Biotin) [clone: KRT19/799 KRT19/800]
Przeciwciała
This antibody recognizes a protein of 40 kDa, identified as cytokeratin-19 (CK19), which is expressed in sweat gland, mammary gland ductal and secretory cells, bile ducts, gastrointestinal tract, bladder urothelium, oral epithelia, esophagus, and ectocervical epithelium. Anti-CK19 reacts with a wide variety of epithelial malignancies including adenocarcinomas of the colon, stomach, pancreas, biliary tract, liver, and breast. Perhaps the most useful application is the identification of thyroid carcinoma of the papillary type, although 50%-60% of follicular carcinomas are also labeled. Anti-CK19 is a useful marker for detection of tumor cells in lymph nodes, peripheral blood, bone marrow and breast cancer.
Type: Primary
Antigen: KRT19
Clonality: Monoclonal
Clone: KRT19/799 KRT19/800
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human, Mouse