Drukuj

Anti-KRT19 Mouse Monoclonal Antibody [clone: KRT19/799 KRT19/800]

Dostawca: Biotium
BNUB1150-500 BNUB1150-100
BNUB1150-500EA 2610 PLN
BNUB1150-500 BNUB1150-100
Anti-KRT19 Mouse Monoclonal Antibody [clone: KRT19/799 KRT19/800]
Przeciwciała
This antibody recognizes a protein of 40 kDa, identified as cytokeratin-19 (CK19), which is expressed in sweat gland, mammary gland ductal and secretory cells, bile ducts, gastrointestinal tract, bladder urothelium, oral epithelia, esophagus, and ectocervical epithelium. Anti-CK19 reacts with a wide variety of epithelial malignancies including adenocarcinomas of the colon, stomach, pancreas, biliary tract, liver, and breast. Perhaps the most useful application is the identification of thyroid carcinoma of the papillary type, although 50%-60% of follicular carcinomas are also labeled. Anti-CK19 is a useful marker for detection of tumor cells in lymph nodes, peripheral blood, bone marrow and breast cancer.

  • Purified

Type: Primary
Antigen: KRT19
Clonality: Monoclonal
Clone: KRT19/799 KRT19/800
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human, Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR