Drukuj

Anti-ZAP70 Mouse Monoclonal Antibody (CF568) [clone: ZAP70/528 + 2F3.2]

Dostawca: Biotium
BNC681137-500 BNC681137-100
BNC681137-500EA 2860 PLN
BNC681137-500 BNC681137-100
Anti-ZAP70 Mouse Monoclonal Antibody (CF568) [clone: ZAP70/528 + 2F3.2]
Przeciwciała
ZAP70 is a 70 kDa protein tyrosine kinase found in T-cells and natural killer cells.Control of this protein translation is via the IgVH gene. ZAP70 protein is expressed in leukemic cells of approximately 25% of chronic lymphocytic leukemia (CLL) cases as well.Anti-ZAP70 expression is an excellent surrogate marker for the distinction between the Ig-mutated (anti-ZAP70 negative) and Ig-unmutated (anti-ZAP70 positive) CLL subtypes and can identify patient groups with divergent clinical courses. The anti-ZAP70 positive Ig-unmutated CLL cases have been shown to have a poorer prognosis.

Type: Primary
Antigen: ZAP70
Clonality: Monoclonal
Clone: ZAP70/528 + 2F3.2
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR