Drukuj

Anti-MCAM Mouse Monoclonal Antibody (CF640R) [clone: MUC18/1130]

Dostawca: Biotium
BNC401130-500 BNC401130-100
BNC401130-500EA 2860 PLN
BNC401130-500 BNC401130-100
Anti-MCAM Mouse Monoclonal Antibody (CF640R) [clone: MUC18/1130]
Przeciwciała
The human Mel-CAM gene maps to chromosome 11q23 and encodes a trans-membrane glycoprotein, also designated MCAM, MUC 18 or CD146, that belongs to the immunoglobulin superfamily and functions as a Ca2 -independent cell adhesion molecule. Mel-CAM expression is restricted to advanced primary and metastatic melanomas and to cell lines of the neuroectodermal lineage, but not normal melanocytes. Mel-CAM is found on 80% of advanced primary human melanomas and correlates well with development of metastatic disease.

Type: Primary
Antigen: MCAM
Clonality: Monoclonal
Clone: MUC18/1130
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR