Drukuj
Anti-FSHB Mouse Monoclonal Antibody (CF568) [clone: FSHb/1062]
Dostawca: Biotium
BNC681062-500 BNC681062-100
BNC681062-500EA 2860 PLN
BNC681062-500 BNC681062-100
Anti-FSHB Mouse Monoclonal Antibody (CF568) [clone: FSHb/1062]
Przeciwciała
This MAb reacts with a protein of 22 kDa, identified as beta sub-unit of FSH. It does not cross react with the alpha sub-unit. Follicle stimulating hormone (FSH) is a hormone synthesized and secreted by gonadotrophs in the anterior pituitary gland. In the ovary, FSH stimulates the growth of immature Graafian follicles to maturation. As the follicle grows, it releases inhibin, which deactivates the FSH production. In men, FSH enhances the production of androgen-binding protein by the Sertoli cells of the testis and is critical for spermatogenesis. FSH and LH act synergistically in reproduction. FSH is a useful marker in the classification of pituitary tumors and the study of pituitary disease.
Type: Primary
Antigen: FSHB
Clonality: Monoclonal
Clone: FSHb/1062
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human