Drukuj

Anti-CHGA Mouse Monoclonal Antibody [clone: CGA414 CGA/777 CGA/798]

Dostawca: Biotium
BNUB1060-500 BNUB1060-100
BNUB1060-500EA 2610 PLN
BNUB1060-500 BNUB1060-100
Anti-CHGA Mouse Monoclonal Antibody [clone: CGA414 CGA/777 CGA/798]
Przeciwciała
Chromogranin A is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, pheochromocytomas, paragangliomas, and other neuroendocrine tumors. Co-expression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms. Reportedly, co-expression of certain keratins and chromogranin indicates neuroendocrine lineage. The presence of strong anti-chromogranin staining and absence of anti-keratin staining should raise the possibility of paraganglioma. The co-expression of chromogranin and NSE is typical of neuroendocrine neoplasms. Most pituitary adenomas and prolactinomas readily express chromogranin.

  • Purified

Type: Primary
Antigen: CHGA
Clonality: Monoclonal
Clone: CGA414 CGA/777 CGA/798
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human, Monkey, Mouse, Pig, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR