Drukuj

Anti-CD84 Mouse Monoclonal Antibody (CF405S) [clone: 153-4D9]

Dostawca: Biotium
BNC041049-500 BNC041049-100
BNC041049-500EA 2860 PLN
BNC041049-500 BNC041049-100
Anti-CD84 Mouse Monoclonal Antibody (CF405S) [clone: 153-4D9]
Przeciwciała
Recognizes a protein of 74 kDa, identified as CD84. It is expressed on mature B cells and on B-cell lines, including pre-B-cell lines, but not on plasma cell lines. Immunohistochemical studies demonstrated that CD84 strongly expressed on tissues macrophages. CD84 is also highly expressed on platelets and, at low levels on peripheral blood T cells. It is a highly N-glycosylated protein and belongs to immunoglobulin superfamily. It may play a role in leukocyte activation.

Type: Primary
Antigen: CD84
Clonality: Monoclonal
Clone: 153-4D9
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR