Drukuj

Anti-S100A1 Mouse Monoclonal Antibody (CF568) [clone: S100A1/1012]

Dostawca: Biotium
BNC681012-500 BNC681012-100
BNC681012-500EA 2860 PLN
BNC681012-500 BNC681012-100
Anti-S100A1 Mouse Monoclonal Antibody (CF568) [clone: S100A1/1012]
Przeciwciała
The family of EF-hand type Ca2 -binding proteins includes calbindin, S-100 alpha and beta, calgranulins, B and C, and the parvalbumin family members, including parvalbumin alpha and parvalbumin beta. The S-100 protein is involved in the regulation of cellular processes such as cell cycle progression and differentiation. S-100 protein may function in the activation of Ca2 induced Ca2 release, inhibition of microtubule assembly and inhibition of protein kinase C mediated phosphorylation. Two S-100 subunits, sharing 60% sequence identity, have been described as S-100 alpha chain and S-100 beta chain. Three S-100 dimeric forms have been characterized, differing in their subunit composition of two alpha chains, two beta chains or one alpha and one beta chain. S-100 localizes to the cytoplasm and nuclei of astrocytes, Schwann s cells, ependymomas and astrogliomas. S-100 is also detected in almost all benign naevi, malignant melanocytic tumours and in Langerhans cells in the skin. Calbindin, S-100 proteins and parvalbumin proteins are each expressed in neural tissues. In addition, S-100 alpha and beta are present in a variety of other tissues and calbindin is present in intestine and kidney.

Type: Primary
Antigen: S100A1
Clonality: Monoclonal
Clone: S100A1/1012
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Cow, Human, Mouse, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR