Drukuj

Anti-B2M Mouse Monoclonal Antibody (CF405S) [clone: B2M/961]

Dostawca: Biotium
BNC040961-500 BNC040961-100
BNC040961-500EA 2860 PLN
BNC040961-500 BNC040961-100
Anti-B2M Mouse Monoclonal Antibody (CF405S) [clone: B2M/961]
Przeciwciała
Recognizes a protein of 12 kDa, identified as β-2 microglobulin. Major histocompatibility complex (MHC) class 1 molecules bind to antigens for presentation on the surface of cells. The proteasome is responsible for producing these antigens from the components of foreign pathogens. MHC class 1 molecules consist of an α heavy chain that contains three subdomains (α1, α2, α3) and a non-covalent associating light chain, known as β-2-Microglobulin. β-2-Microglobulin associates with the α3 subdomain of the α heavy chain and forms an immunoglobulin domain-like structure that mediates proper folding and expression of MHC class 1 molecules. The α1 and α2 domains of the α heavy chain form the peptide antigen-binding cleft. Mutations in the β-2-Microglobulin gene can enhance the progression of malignant melanoma phenotypes.

Type: Primary
Antigen: B2M
Clonality: Monoclonal
Clone: B2M/961
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Non-human primates
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR