Drukuj
Anti-IGHG Mouse Monoclonal Antibody (Purified) [clone: B33/20]
Dostawca: Biotium
BNUB0952-500 BNUB0952-100
BTIUBNUB0952-500EA 2610 PLN
BTIUBNUB0952-500 BTIUBNUB0952-100
Anti-IGHG Mouse Monoclonal Antibody (Purified) [clone: B33/20]
Przeciwciała
Recognizes a protein of 75 kDa, identified as γ heavy chain of human immunoglobulins. Its epitope maps in CH2 domain of Fc region of IgG. It reacts with all sub-classes of γ chain of human immunoglobulins. It does not cross-react with α (IgA), μ (IgM), ε (IgE), or δ (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. This MAb is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. The most common feature of these malignancies is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.
Type: Primary
Antigen: IGHG
Clonality: Monoclonal
Clone: B33/20
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human