Drukuj

Anti-CD68 Mouse Monoclonal Antibody (Purified) [clone: CD68/G2]

Dostawca: Biotium
BNUB0919-500 BNUB0919-100
BNUB0919-500EA 2610 PLN
BNUB0919-500 BNUB0919-100
Anti-CD68 Mouse Monoclonal Antibody (Purified) [clone: CD68/G2]
Przeciwciała
This antibody recognizes a glycoprotein of 110 kDa, which is identified as CD68. It is important for identifying macrophages in tissue sections. It stains macrophages in a wide variety of human tissues, including Kupffer cells and macrophages in the red pulp of the spleen, in lamina propria of the gut, in lung alveoli, and in bone marrow. It reacts with myeloid precursors and peripheral blood granulocytes. It also reacts with plasmacytoid T cells, which are supposed to be of monocyte/macrophage origin. It shows strong granular cytoplasmic staining of chronic and acute myeloid leukemia and also reacts with rare cases of true histiocytic neoplasia. Lymphomas are negative or show few granules.

Type: Primary
Antigen: CD68
Clonality: Monoclonal
Clone: CD68/G2
Conjugation: Purified
Epitope:
Host: Mouse
Reactivity: Human
Isotype: IgG1, kappa
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR