Drukuj

Anti-HSPD1 Mouse Monoclonal Antibody (CF405S) [clone: HSPD1/875]

Dostawca: Biotium
BNC040875-500 BNC040875-100
BNC040875-500EA 2860 PLN
BNC040875-500 BNC040875-100
Anti-HSPD1 Mouse Monoclonal Antibody (CF405S) [clone: HSPD1/875]
Przeciwciała
Recognizes a 60 kDa protein, identified as the heat shock protein 60 (hsp60). A wide variety of environmental and pathophysiological stressful conditions trigger the synthesis of a family of proteins known as heat shock proteins (hsp), more appropriately called as stress response proteins (srp). hsp60 is a potential antigen in a number of autoimmune diseases. In human arthritis and in experimentally induced arthritis in animals, disease development coincides with the development of immune reactivity directed against not only bacterial hsp60, but also against its mammalian homolog.

Type: Primary
Antigen: HSPD1
Clonality: Monoclonal
Clone: HSPD1/875
Conjugation: CF405S
Epitope:
Host: Mouse
Reactivity: Chicken, Guinea Pig, Hamster, Human, Monkey, Mouse, Pig, Rat
Isotype: IgG1, kappa
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR