Drukuj

Anti-RBP1 Mouse Monoclonal Antibody (CF488A) [clone: G4E4]

Dostawca: Biotium
BNC880855-500 BNC880855-100
BNC880855-500EA 2860 PLN
BNC880855-500 BNC880855-100
Anti-RBP1 Mouse Monoclonal Antibody (CF488A) [clone: G4E4]
Przeciwciała
This MAb recognizes an epitope within the 74-182 C-terminal sequence (11kD peptide fragment) of human serum Cellular Retinol Binding Protein 1 (CRBP 1), a single-chain glycoprotein belonging to the superfamily of hydrophobic molecule transporter proteins, which is responsible for transport of retinol (vitamin A1) from the liver to peripheral target tissues, like the eye, where it mediates the cellular uptake. CRBP 1 is synthesized by hepatic parenchymal cells where it becomes bound to its ligand retinol and is then released into the circulation, where it binds further to the protein transthyretin, to form a transporting complex, which is big enough not to be lost by filtration through the kidney glomeruli. It is detected in nearly all tissues with higher expression in adult ovary, pancreas, pituitary gland, adrenal gland, and fetal liver.

Type: Primary
Antigen: RBP1
Clonality: Monoclonal
Clone: G4E4
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Chimpanzee, Goat, Human, Monkey, Mouse, Rabbit, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR