Drukuj
Anti-MUC2 Mouse Monoclonal Antibody (CF568) [clone: MLP/842]
Dostawca: Biotium
BNC680842-500 BNC680842-100
BNC680842-500EA 2860 PLN
BNC680842-500 BNC680842-100
Anti-MUC2 Mouse Monoclonal Antibody (CF568) [clone: MLP/842]
Przeciwciała
Recognizes a single glycoprotein of 520 kDa, identified as mucin 2 (MUC2). This MAb shows no cross-reaction with human milk fat globule membranes, MUC1, or MUC3. Mucins are high molecular weight glycoproteins, which constitute the major component of the mucus layer that protects the gastric epithelium.MUC2 is specifically expressed in goblet cells of the small intestine & colon; in about 65% of colonic carcinomas and about 40% of gastric carcinomas. MUC2 is rarely expressed outside of the GI tract with the exceptions of mucinous carcinoma of breast and clear cell-type carcinomas of the ovary.
Type: Primary
Antigen: MUC2
Clonality: Monoclonal
Clone: MLP/842
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human