Drukuj

Anti-CD79A Mouse Monoclonal Antibody (CF640R) [clone: JCB117 HM47/A9]

Dostawca: Biotium
BNC400828-500 BNC400828-100
BNC400828-500EA 2860 PLN
BNC400828-500 BNC400828-100
Anti-CD79A Mouse Monoclonal Antibody (CF640R) [clone: JCB117 HM47/A9]
Przeciwciała
A disulphide-linked heterodimer, consisting of mb-1 (or CD79a) and B29 (or CD79b) polypeptides, is non-covalently associated with membrane-bound immunoglobulins on B cells. This complex of mb-1 and B29 polypeptides and immunoglobulin constitute the B cell Ag receptor. CD79a first appears at pre B cell stage, early in maturation, and persists until the plasma cell stage where it is found as an intracellular component. CD79a is found in the majority of acute leukemias of precursor B cell type, in B cell lines, B cell lymphomas, and in some myelomas. It is not present in myeloid or T cell lines. Anti-CD79a is generally used to complement anti-CD20 especially for mature B-cell lymphomas after treatment with Rituximab (anti-CD20). This antibody will stain many of the same lymphomas as anti-CD20, but also is more likely to stain B-lymphoblastic lymphoma/leukemia than is anti-CD20. Anti-CD79a also stains more cases of plasma cell myeloma and occasionally some types of endothelial cells as well.

Type: Primary
Antigen: CD79A
Clonality: Monoclonal
Clone: JCB117 HM47/A9
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Cow, Human, Monkey, Mouse, Pig, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR