Drukuj
Anti-CALD1 Mouse Monoclonal Antibody (CF594) [clone: CALD1/820]
Dostawca: Biotium
BNC940820-500 BNC940820-100
BNC940820-500EA 2860 PLN
BNC940820-500 BNC940820-100
Anti-CALD1 Mouse Monoclonal Antibody (CF594) [clone: CALD1/820]
Przeciwciała
Recognizes a protein of 150 kDa, which is identified as the high molecular weight variant of Caldesmon. Two closely related variants of human caldesmon have been identified which are different in their electrophoretic mobility and cellular distribution. The h-caldesmon variant (120-150 kDa) is predominantly expressed in smooth muscle whereas l-caldesmon (70-80 kDa) is found in non- muscle tissue and cells. Neither of the two variants has been detected in skeletal muscle. This MAb recognizes only the 150 kDa variant (h-caldesmon) in Western blots of human aortic media extracts and is unreactive with fibroblast extracts from cultivated human foreskin. Caldesmon is a developmentally regulated protein involved in smooth muscle and non-muscle contraction.
Type: Primary
Antigen: CALD1
Clonality: Monoclonal
Clone: CALD1/820
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human