Drukuj

Anti-ACTA2 Mouse Monoclonal Antibody (CF594) [clone: ACTA2/791]

Dostawca: Biotium
BNC940791-500 BNC940791-100
BNC940791-500EA 2860 PLN
BNC940791-500 BNC940791-100
Anti-ACTA2 Mouse Monoclonal Antibody (CF594) [clone: ACTA2/791]
Przeciwciała
Actin is a major component of the cytoskeleton and is present in most cell types. It is highly specific to actin from smooth muscles. This MAb does not stain cardiac or skeletal muscle; however, it does stain myofibroblasts and myoepithelial cells. This antibody could be used together with anti-muscle specific actin and myogenin in making a diagnosis of smooth muscle and skeletal muscle tumors. In most cases of rhabdomyosarcoma, this antibody yields negative results whereas anti-muscle specific actin and myogenin are positive. Leiomyosarcomas are positive only with anti-muscle specific actin and anti-smooth muscle actin and are negative with anti-myogenin.

Type: Primary
Antigen: ACTA2
Clonality: Monoclonal
Clone: ACTA2/791
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR