Drukuj
Anti-HSPD1 Mouse Monoclonal Antibody (Biotin) [clone: HSPD1/780]
Dostawca: Biotium
BNCB0780-500 BNCB0780-100
BTIUBNCB0780-500EA 2860 PLN
BTIUBNCB0780-500 BTIUBNCB0780-100
Anti-HSPD1 Mouse Monoclonal Antibody (Biotin) [clone: HSPD1/780]
Przeciwciała
Recognizes a 60 kDa protein, identified as the heat shock protein 60 (hsp60). A wide variety of environmental and pathophysiological stressful conditions trigger the synthesis of a family of proteins known as heat shock proteins (hsp), more appropriately called as stress response proteins (srp). Hsp60 is a potential antigen in a number of autoimmune diseases. In human arthritis and in experimentally induced arthritis in animals, disease development coincides with the development of immune reactivity directed against not only bacterial hsp60, but also against its mammalian homolog.
Type: Primary
Antigen: HSPD1
Clonality: Monoclonal
Clone: HSPD1/780
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Chicken, Cow, Dog, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep