Drukuj

Anti-IGHM Mouse Monoclonal Antibody (CF647) [clone: DA4-4]

Dostawca: Biotium
BNC470759-500 BNC470759-100
BNC470759-500EA 2860 PLN
BNC470759-500 BNC470759-100
Anti-IGHM Mouse Monoclonal Antibody (CF647) [clone: DA4-4]
Przeciwciała
Recognizes a protein of 75 kDa, identified as mu heavy chain of human immunoglobulins. It does not cross-react with alpha (IgA), gamma (IgG), epsilon (IgE), or delta (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. This MAb is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. The most common feature of these malignancies is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.

Type: Primary
Antigen: IGHM
Clonality: Monoclonal
Clone: DA4-4
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR