Drukuj
Anti-KRT19 Mouse Monoclonal Antibody [clone: A53-B/A2.26 BA17]
Dostawca: Biotium
BNUB0753-500 BNUB0753-100
BNUB0753-500EA 2610 PLN
BNUB0753-500 BNUB0753-100
Anti-KRT19 Mouse Monoclonal Antibody [clone: A53-B/A2.26 BA17]
Przeciwciała
This MAb reacts with the rod domain of human cytokeratin-19 (CK19), a polypeptide of 40 kDa. CK19 is expressed in sweat gland, mammary gland ductal and secretory cells, bile ducts, gastrointestinal tract, bladder urothelium, oral epithelia, esophagus, and ectocervical epithelium. Anti-CK19 reacts with a wide variety of epithelial malignancies including adenocarcinomas of the colon, stomach, pancreas, biliary tract, liver, and breast. Perhaps the most useful application is the identification of thyroid carcinoma of the papillary type, although 50%-60% of follicular carcinomas are also labeled. Anti-CK19 is a useful marker for detection of tumor cells in lymph nodes, peripheral blood, bone marrow and breast cancer.
  • Purified
Type: Primary
Antigen: KRT19
Clonality: Monoclonal
Clone: A53-B/A2.26 BA17
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human