Drukuj
Anti-PRLR Mouse Monoclonal Antibody (CF488A) [clone: B6.2 PRLR742]
Dostawca: Biotium
BNC880744-500 BNC880744-100
BNC880744-500EA 2860 PLN
BNC880744-500 BNC880744-100
Anti-PRLR Mouse Monoclonal Antibody (CF488A) [clone: B6.2 PRLR742]
Przeciwciała
This antibody recognizes a protein of 70 kDa, identified as prolactin receptor. Prolactin is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. The initial step in its action is the binding to a specific membrane receptor (prolactin receptor), which belongs to the superfamily of class 1 cytokine receptors. The function of the prolactin receptor is mediated, at least in part, by two families of signaling molecules: Janus kinases and signal transducers and activators of transcription.
Type: Primary
Antigen: PRLR
Clonality: Monoclonal
Clone: B6.2 PRLR742
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human