Drukuj

Anti-PCNA Mouse Monoclonal Antibody (CF405S) [clone: PCNA/694]

Dostawca: Biotium
BNC040694-500 BNC040694-100
BNC040694-500EA 2860 PLN
BNC040694-500 BNC040694-100
Anti-PCNA Mouse Monoclonal Antibody (CF405S) [clone: PCNA/694]
Przeciwciała
Recognizes a non-histone protein of 36 kDa, which is identified as proliferating cell nuclear antigen (PCNA). It is also known as cyclin or polymerase delta auxiliary protein. Elevated expression of PCNA/cyclin has been shown in the nucleus during late G1 phase immediately before the onset of DNA synthesis, becoming maximal during S-phase and declining during G2 and M phases. This MAb is excellent for multiple applications.

Type: Primary
Antigen: PCNA
Clonality: Monoclonal
Clone: PCNA/694
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR