Drukuj
Anti-CD68 Mouse Monoclonal Antibody (CF568) [clone: C68/684 KP1]
Dostawca: Biotium
BNC680685-500 BNC680685-100
BNC680685-500EA 2860 PLN
BNC680685-500 BNC680685-100
Anti-CD68 Mouse Monoclonal Antibody (CF568) [clone: C68/684 KP1]
Przeciwciała
This antibody recognizes a glycoprotein of 110 kDa, which is identified as CD68. It is important for identifying macrophages in tissue sections. It stains macrophages in a wide variety of human tissues, including Kupffer cells and macrophages in the red pulp of the spleen, in lamina propria of the gut, in lung alveoli, and in bone marrow. It reacts with myeloid precursors and peripheral blood granulocytes. It also reacts with plasmacytoid T cells, which are supposed to be of monocyte/macrophage origin. It shows strong granular cytoplasmic staining of chronic and acute myeloid leukemia and also reacts with rare cases of true histiocytic neoplasia. Lymphomas are negative or show few granules.
Type: Primary
Antigen: CD68
Clonality: Monoclonal
Clone: Clone C68/684 KP1
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Cat, Human, Monkey