Drukuj

Anti-Hepatocyte Specific Mouse Monoclonal Antibody [clone: OCH1E5]

Dostawca: Biotium
BNUB0671-500 BNUB0671-100
BNUB0671-500EA 2610 PLN
BNUB0671-500 BNUB0671-100
Anti-Hepatocyte Specific Mouse Monoclonal Antibody [clone: OCH1E5]
Przeciwciała
Hepatocyte Specific Antigen, also called Hepatocyte Paraffin 1 or HepPar1, localizes to the mitochondria of hepatocytes. It is a sensitive marker for distinguishing hepatocellular carcinomas (HCC) from other metastatic carcinomas as well as cholangio-carcinomas. HCC's occur primarily in the stomach, but they are also found in many other organs. The Hepatocyte Specific Antigen may also be a useful marker for intestinal metaplasia. Reportedly, strong expression of the Hepatocyte Specific Antigen correlates with smaller tumor size and longer patient survival. Occasionally, Hepatocyte Specific Antigen is also found in gastric carcinomas as well as in a few other non-hepatic tumors.

  • Purified

Type: Primary
Antigen: Hepatocyte Specific
Clonality: Monoclonal
Clone: OCH1E5
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Dog, Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR