Drukuj

Anti-CHGA Mouse Monoclonal Antibody (CF568) [clone: LK2H10]

Dostawca: Biotium
BNC680641-500 BNC680641-100
BNC680641-500EA 2860 PLN
BNC680641-500 BNC680641-100
Anti-CHGA Mouse Monoclonal Antibody (CF568) [clone: LK2H10]
Przeciwciała
Chromogranin A is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, pheochromocytomas, paragangliomas, and other neuroendocrine tumors. Co-expression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms. Reportedly, co-expression of certain keratins and chromogranin indicates neuroendocrine lineage. The presence of strong anti-chromogranin staining and absence of anti-keratin staining should raise the possibility of paraganglioma. The co-expression of chromogranin and NSE is typical of neuroendocrine neoplasms. Most pituitary adenomas and prolactinomas readily express chromogranin.

Type: Primary
Antigen: CHGA
Clonality: Monoclonal
Clone: LK2H10
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Monkey, Mouse, Pig, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR