Drukuj
Anti-VWF Mouse Monoclonal Antibody (CF555) [clone: 3E2D10]
Dostawca: Biotium
BNC550633-500 BNC550633-100
BNC550633-500EA 2860 PLN
BNC550633-500 BNC550633-100
Anti-VWF Mouse Monoclonal Antibody (CF555) [clone: 3E2D10]
Przeciwciała
von Willebrand Factor (vWF) is a multimeric glycoprotein that is found in endothelial cells, plasma and platelets. It acts as a carrier protein for Factor VIII and promotes platelet adhesion and aggregation. vWF undergoes a variety of posttranslational modifications that influence the affinity and availability for Factor VIII, including cleavage of the propeptide and formation of N-terminal disulfide bonds. This antibody helps to establish the endothelial nature of some lesions of disputed histogenesis, e.g. Kaposi's sarcoma and cardiac myxoma. It is widely used for differentiating vascular lesions from those of other tissue differentiation within a panel of other vascular markers although not all tumors of endothelial differentiation contain this antigen.
Type: Primary
Antigen: VWF
Clonality: Monoclonal
Clone: 3E2D10
Conjugation: CF555
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human