Drukuj
Anti-AFP Mouse Monoclonal Antibody (CF594) [clone: C2]
Dostawca: Biotium
BNC940523-500 BNC940523-100
BNC940523-500EA 2860 PLN
BNC940523-500 BNC940523-100
Anti-AFP Mouse Monoclonal Antibody (CF594) [clone: C2]
Przeciwciała
This antibody recognizes an oncofetal glycoprotein with a single chain of 70 kDa, which is identified as alpha fetoprotein (AFP) (ISOBM TD-2 workshop). This MAb is highly specific to AFP and shows no cross-reaction with other oncofetal antigens or serum albumin. AFP is normally synthesized in the liver, intestinal tract, and yolk sac of the fetus. Antibody to AFP has been shown to be useful in detecting hepatocellular carcinomas (HCC) and germ cell neoplasms, especially yolk sac tumors.
Type: Primary
Antigen: AFP
Clonality: Monoclonal
Clone: C2
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse