Drukuj

Anti-MSN Mouse Monoclonal Antibody (CF640R) [clone: MSN493]

Dostawca: Biotium
BNC400493-500 BNC400493-100
BNC400493-500EA 2860 PLN
BNC400493-500 BNC400493-100
Anti-MSN Mouse Monoclonal Antibody (CF640R) [clone: MSN493]
Przeciwciała
Recognizes 78 kDa moesin protein. Moesin, a member of the talin-4.1 superfamily, is a linking protein of the sub-membranous actin cytoskeleton. It is expressed in variable amounts in cells of different phenotypes such as macrophages, lymphocytes, fibroblastic, endothelial, epithelial, and neuronal cell lines but not in blood cells. The ERM proteins, ezrin, radixin, and moesin are involved in a variety of cellular functions, such as cell adhesion, migration, and the organization of cell surface structures, and are highly homologous, both in protein sequence and in functional activity, with merlin/schwannomin, a neurofibromatosis-2-associated tumor-suppressor protein. Cell lines of epithelial and mesothelial origin contain both moesin and radixin whereas cells of endothelial and lymphoid origin express moesin.

Type: Primary
Antigen: MSN
Clonality: Monoclonal
Clone: MSN493
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR