Drukuj

Anti-KRT19 Mouse Monoclonal Antibody (CF640R) [clone: Ks19.1]

Dostawca: Biotium
BNC400394-500 BNC400394-100
BNC400394-500EA 2860 PLN
BNC400394-500 BNC400394-100
Anti-KRT19 Mouse Monoclonal Antibody (CF640R) [clone: Ks19.1]
Przeciwciała
This MAb reacts with the rod domain of human cytokeratin-19 (CK19), a polypeptide of 40 kDa. Its epitope maps between amino acid 312-335. CK19 is expressed in sweat gland, mammary gland ductal and secretory cells, bile ducts, gastrointestinal tract, bladder urothelium, oral epithelia, esophagus, and ectocervical epithelium. Anti-CK19 reacts with a wide variety of epithelial malignancies including adenocarcinomas of the colon, stomach, pancreas, biliary tract, liver, and breast. Perhaps the most useful application is the identification of thyroid carcinoma of the papillary type, although 50%-60% of follicular carcinomas are also labeled. Anti-CK19 is a useful marker for detection of tumor cells in lymph nodes, peripheral blood, bone marrow and breast cancer.

Type: Primary
Antigen: KRT19
Clonality: Monoclonal
Clone: Ks19.1
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR