Drukuj
Anti-TNC Mouse Monoclonal Antibody (Biotin) [clone: T2H5]
Dostawca: Biotium
BNCB0392-500 BNCB0392-100
BNCB0392-500EA 2860 PLN
BNCB0392-500 BNCB0392-100
Anti-TNC Mouse Monoclonal Antibody (Biotin) [clone: T2H5]
Przeciwciała
In Western blotting, this antibody reacts with two bands of ~MW of 210 kDa and 300 kDa, identified as two isoforms of Tenascin C. Specificity of this MAb is validated by sequential immunoprecipitation with a PAb against Tenascin C. Tenascin C is a multifunctional, disulfide-linkedhexameric extracellular matrix glycoprotein expressed in association with mesenchymal epithelial interactions during development and in the neo-vasculature and stroma of undifferentiated tumors. In adults, it is restricted to certain epithelial-stromal interfaces and increases markedly in hyper-proliferative diseases and in stroma of many neoplasms, including gliomas, breast, squamous and lung carcinomas.
Type: Primary
Antigen: TNC
Clonality: Monoclonal
Clone: T2H5
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human