Drukuj

Anti-ERBB2 Mouse Monoclonal Antibody (Purified) [clone: HRB2/273]

Dostawca: Biotium
BNUB0273-500 BNUB0273-100
BNUB0273-500EA 2610 PLN
BNUB0273-500 BNUB0273-100
Anti-ERBB2 Mouse Monoclonal Antibody (Purified) [clone: HRB2/273]
Przeciwciała
This MAb is specific to c-erbB-2/HER-2 and shows minimal cross-reaction with other members of the family. C-erbB-2/HER-2 is a member of the EGFR family. Receptors of this family are located on the plasma membrane and consist of an extracellular ligand-binding domain that is connected to a large intracellular domain by a single transmembrane sequence. c-erbB-2/HER-2 protein is over-expressed in a variety of carcinomas especially those of breast and ovary.

Type: Primary
Antigen: ERBB2
Clonality: Monoclonal
Clone: HRB2/273
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR