Drukuj
Anti-ERBB2 Mouse Monoclonal Antibody (CF568) [clone: HRB2/258]
Dostawca: Biotium
BNC680258-500 BNC680258-100
BNC680258-500EA 2860 PLN
BNC680258-500 BNC680258-100
Anti-ERBB2 Mouse Monoclonal Antibody (CF568) [clone: HRB2/258]
Przeciwciała
Recognizes a protein of 185 kDa, which is identified as c-erbB-2/HER-2/neu. Its epitope is localized in the extracellular domain. C-erbB-2/HER-2 is a member of the EGFR family. This MAb is specific and shows minimal cross-reaction with other members of the EGFR-family. Receptors of this family are located on the plasma membrane and consist of an extracellular ligand-binding domain that is connected to a large intracellular domain by a single transmembrane sequence. C-erbB-2/HER-2 protein is over-expressed in a variety of carcinomas especially those of breast and ovary.
Type: Primary
Antigen: ERBB2
Clonality: Monoclonal
Clone: HRB2/258
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Rat