Drukuj

Anti-ALPP Mouse Monoclonal Antibody (CF640R) [clone: ALPP/238]

Dostawca: Biotium
BNC400238-500 BNC400238-100
BNC400238-500EA 2860 PLN
BNC400238-500 BNC400238-100
Anti-ALPP Mouse Monoclonal Antibody (CF640R) [clone: ALPP/238]
Przeciwciała
PLAP is a tissue specific, trophoblast-derived, 70 kDa, glycosyl-phosphatidylinositol (GPI)-anchored, dimeric, Zn2 metallo glycoprotein that catalyzes the hydrolysis of phosphomonoesters into an inorganic phosphate and an alcohol. It is present in the placenta and serum of pregnant women and in high frequency in gynecological and testicular cancers and in lower frequency in other tumors. The three tissue-specific AP's in humans, PLAP, germ cell AP (GCAP) and intestinal AP, are 90-98% homologous. Non-tissue specific AP is found in kidney, liver and bone. This MAb binds equally well to all common allelic variants (S, F, FS and I) of PLAP as to AP from normal human testis. This MAb can be used both as coating as well as tracer antibody in the same ELISA to detect PLAP in serum of S, F, FS and I phenotypes.

Type: Primary
Antigen: ALPP
Clonality: Monoclonal
Clone: ALPP/238
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR