Drukuj
Anti-KRT18 Mouse Monoclonal Antibody (CF640R) [clone: DA7]
Dostawca: Biotium
BNC400198-500 BNC400198-100
BNC400198-500EA 2860 PLN
BNC400198-500 BNC400198-100
Anti-KRT18 Mouse Monoclonal Antibody (CF640R) [clone: DA7]
Przeciwciała
This MAb reacts with a wide variety of simple epithelia. It does not react with stratified squamous epithelia. It reacts with epithelial tumors of the gastrointestinal tract, lung, breast, pancreas, ovary, and thyroid. Cytokeratin 18, which belongs to the type A (acidic) subfamily of low molecular weight keratins, exists in combination with cytokeratin 8. It was reported that tissues from gastrointestinal tract are positive for both cytokeratin 8 and 18 but do not contain cytokeratin 14. Tissues from gastrointestinal tract, respiratory tract and urogenital tract, as well as endocrine and exocrine tissues and mesothelial cells are positive for cytokeratin 18.
Type: Primary
Antigen: KRT18
Clonality: Monoclonal
Clone: DA7
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human