Drukuj

Anti-GFAP Mouse Monoclonal Antibody (CF568) [clone: GA-5]

Dostawca: Biotium
BNC680167-500 BNC680167-100
BNC680167-500EA 2860 PLN
BNC680167-500 BNC680167-100
Anti-GFAP Mouse Monoclonal Antibody (CF568) [clone: GA-5]
Przeciwciała
This MAb recognizes a protein of ~50 kDa which is identified as Glial Fibrillary Acidic Protein (GFAP). It shows no cross-reaction with other intermediate filament proteins. GFAP is specifically found in astroglia. GFAP is a very popular marker for localizing benign astrocyte and neoplastic cells of glial origin in the central nervous system. Antibody to GFAP is useful in differentiating primary gliomas from metastatic lesions in the brain and for documenting astrocytic differentiation in tumors outside the CNS.

Type: Primary
Antigen: GFAP
Clonality: Monoclonal
Clone: GA-5
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Cow, Human, Mouse, Pig, Rabbit, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR