Drukuj

Anti-ITGAL Mouse Monoclonal Antibody (CF568) [clone: Cris-3]

Dostawca: Biotium
BNC680151-500 BNC680151-100
BNC680151-500EA 2860 PLN
BNC680151-500 BNC680151-100
Anti-ITGAL Mouse Monoclonal Antibody (CF568) [clone: Cris-3]
Przeciwciała
Recognizes a protein of 180 kDa, identified as CD11a (Leucocyte Workshop II; Code N202). CD11a complex with the 2 subunit of the integrin family, CD18, to form the cell surface heterodimer, LFA-1 or CD11a /C18 (aLbL). LFA-1 is expressed on all leukocytes including lymphocytes, monocytes, and granulocytes. It is involved in leukocyte adhesion to its ligands including intercellular adhesion molecule-1 (ICAM-1 or CD54), ICAM-2 (CD102), ICAM-3 (CD50) and Telencephalin (TLN) and play a role in most immune/inflammatory responses.

Type: Primary
Antigen: ITGAL
Clonality: Monoclonal
Clone: Cris-3
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR