Drukuj
Anti-Human Nuclear Antigen Mouse Monoclonal Antibody (CF543) [clone: NM106]
Dostawca: Biotium
BNC430106-500 BNC430106-100
BNC430106-500EA 2860 PLN
BNC430106-500 BNC430106-100
Anti-Human Nuclear Antigen Mouse Monoclonal Antibody (CF543) [clone: NM106]
Przeciwciała
This MAb is an excellent marker for all nuclei in cells in tissues. It is a part of a new panel of reagents, which recognizes subcellular organelles or compartments of cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. This MAb recognizes an antigen associated with the nuclei in all cells. It can be used to stain the nuclei in cell or tissue preparations and can be used as a nuclear marker in subcellular fractions. It produces a speckled pattern in normal and malignant cells and may be used to stain the nuclei of cells in fixed or frozen tissue sections. It can also be used with paraformaldehyde fixed frozen tissue or cell preparations.
Type: Primary
Antigen: Human Nuclear Antigen
Clonality: Monoclonal
Clone: NM106
Conjugation: CF543
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat