Drukuj

Anti-MLANA Mouse Monoclonal Antibody (Purified) [clone: M2-7C10]

Dostawca: Biotium
BNUB0009-500 BNUB0009-100
BNUB0009-500EA 2610 PLN
BNUB0009-500 BNUB0009-100
Anti-MLANA Mouse Monoclonal Antibody (Purified) [clone: M2-7C10]
Przeciwciała
This antibody recognizes a protein doublet of 20-22 kDa, identified as MART-1 (Melanoma Antigen Recognized by T cells 1) or Melan-A. MART-1 is a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. Seven other melanoma associated antigens recognized by autologous cytotoxic T cells include MAGE-1, MAGE-3, tyrosinase, gp100, gp75, BAGE-1, and GAGE-1. Subcellular fractionation shows that MART-1 is present in melanosomes and endoplasmic reticulum. This MAb labels melanomas and other tumors showing melanocytic differentiation. It is also a useful positive-marker for angiomyolipomas. It does not stain tumor cells of epithelial, lymphoid, glial, or mesenchymal origin.

Type: Primary
Antigen: MLANA
Clonality: Monoclonal
Clone: M2-7C10
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Horse, Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR